Learn More
Enterprise Technology Solutions Hospeco Gloves,Exam,Latex,8mil,9-2/5"L,Large,100/BX,10BX/CT,NL
SDP
Catalog No. 5000828074
Encompass_Preferred
TOTAL GRIP Latex Powder Free Exam Gloves are perfect for the automotive industry. Natural white color helps show any dirt, grease or grime on your hands while working. Gloves on average are 8 mil thick, meaning you have protection for a longer time frame. Latex, exam-grade gloves are powder-free, double-chlorinated and ambidextrous. They are perfect for use in industries such as automotive, aerospace, corrections, first responders, mortuary, contractor supply, laboratory, oil/gas, painting, plumbing, HVAC and more.
Catalog No. 50-008-28074 Supplier Enterprise Technology Solutions Supplier No. HOSGLL165FL
TOTAL GRIP Latex Powder Free Exam Gloves are perfect for the automotive industry. Natural white color helps show any dirt, grease or grime on your hands while working. Gloves on average are 8 mil thick, meaning you have protection for a longer time frame. Latex, exam-grade gloves are powder-free, double-chlorinated and ambidextrous. They are perfect for use in industries such as automotive, aerospace, corrections, first responders, mortuary, contractor supply, laboratory, oil/gas, painting, plumbing, HVAC and more.