Learn More
Enterprise Technology Solutions Hospeco Gloves,Exam,Nitrile,3 mil,9-1/2"L,Large,200/BX,10BX/CT,BVT
SDP
Catalog No. 5000828053
Encompass_Preferred
Nitrile Exam Gloves are made slightly thinner with a 3 mil thickness for ultra-soft, super-flexible comfort, providing more dexterity and tactile sensitivity. Latex-free, nonsterile gloves are standard length with beaded cuffs. They are perfect for patient examinations, dental examinations, general purpose, light-duty industrial applications, foodservice and gardening.
Catalog No. 50-008-28053 Supplier Enterprise Technology Solutions Supplier No. HOSGLN125FL
Nitrile Exam Gloves are made slightly thinner with a 3 mil thickness for ultra-soft, super-flexible comfort, providing more dexterity and tactile sensitivity. Latex-free, nonsterile gloves are standard length with beaded cuffs. They are perfect for patient examinations, dental examinations, general purpose, light-duty industrial applications, foodservice and gardening.