Learn More
Enterprise Technology Solutions Hospeco Gloves,Exam,Nitrile,4 mil,9-1/2"L,XL,100/BX,10BX/CT,BE
SDP
Catalog No. 5000828240
Encompass_Preferred
Nitrile Exam Gloves provide excellent comfort and feel with textured fingers for easy gripping. Cuffs are beaded to prevent gloves from rolling down. They are standard length, latex-free, powder-free and nonsterile. Ambidextrous design is perfect for use in industries such as building services, first responders/corrections, foodservice, government, grocery, healthcare, hospitality and more.
Catalog No. 50-008-28240 Supplier Enterprise Technology Solutions Supplier No. HOSGLN135FX
Nitrile Exam Gloves provide excellent comfort and feel with textured fingers for easy gripping. Cuffs are beaded to prevent gloves from rolling down. They are standard length, latex-free, powder-free and nonsterile. Ambidextrous design is perfect for use in industries such as building services, first responders/corrections, foodservice, government, grocery, healthcare, hospitality and more.