Learn More
Enterprise Technology Solutions Hospeco Gloves,Exam,Nitrile,4 mil,9-2/5"L,XL,90/BX,10BX/CT,BK
SDP
Catalog No. 5000828095
Encompass_Preferred
Night Angel Nitrile Powder Free Exam Gloves fit and feel like latex gloves but are 100 percent free of rubber latex. They are perfect for tattooing, piercing, body art and high-end hair and beauty industries. Dark black color stands out and masks stains, inks and dyes during use, allowing artists to focus on their work. Night Angel gloves help rapidly identify potentially unsafe, light-colored substances and are preferred in law enforcement agencies, correctional facilities, homeland security and postal facilities. Slightly thin design with 4 mil thickness provides an ultra-soft, super-flexible experience for more dexterity and tactile sensitivity.
Catalog No. 50-008-28095 Supplier Enterprise Technology Solutions Supplier No. HOSNGL228
Night Angel Nitrile Powder Free Exam Gloves fit and feel like latex gloves but are 100 percent free of rubber latex. They are perfect for tattooing, piercing, body art and high-end hair and beauty industries. Dark black color stands out and masks stains, inks and dyes during use, allowing artists to focus on their work. Night Angel gloves help rapidly identify potentially unsafe, light-colored substances and are preferred in law enforcement agencies, correctional facilities, homeland security and postal facilities. Slightly thin design with 4 mil thickness provides an ultra-soft, super-flexible experience for more dexterity and tactile sensitivity.