Learn More
Enterprise Technology Solutions Hospeco Gloves,Exam,Nitrile,5.5 mil,9-1/2"L,Small,100/BX,10BX/CT,BE
SDP
Catalog No. 5000828093
Encompass_Preferred
NPF Nitrile Powder Free Exam Gloves are formulated with 100 percent synthetic nitrile polymer and are latex-free, powder-free and odor-free. 5.5 mil thick gloves are softer and more elastic compared to other gloves with the same thickness. Ambidextrous design provides superior resistance to punctures, tears and chemicals. Textured surface allows a strong grip for wet and dry application. They are perfect for use in industries such as automotive, aerospace, building services, first responders, corrections, government, grocery, hospitality and more.
Catalog No. 50-008-28093 Supplier Enterprise Technology Solutions Supplier No. HOSGLN106FS
NPF Nitrile Powder Free Exam Gloves are formulated with 100 percent synthetic nitrile polymer and are latex-free, powder-free and odor-free. 5.5 mil thick gloves are softer and more elastic compared to other gloves with the same thickness. Ambidextrous design provides superior resistance to punctures, tears and chemicals. Textured surface allows a strong grip for wet and dry application. They are perfect for use in industries such as automotive, aerospace, building services, first responders, corrections, government, grocery, hospitality and more.