Learn More
Enterprise Technology Solutions Hospeco Gloves,Exam,Vinyl,4 mil,9-2/5"L,XL,90/BX,10BX/CT,TLT
SDP
Catalog No. 5000828057
Encompass_Preferred
Vinyl Powder Free Exam Gloves provide a latex-free and economical solution to hand protection. Formulated from a special synthetic component and soft, PVC material, these gloves are soft like premium leather and fit snugly and comfortably on your hands. They are 100 percent free of rubber latex, eliminating the risk of developing latex protein-related allergies. 4 mil thick, powder-free gloves are perfect for use in industries such as cosmetology, food-processing, government, grocery, healthcare, hospitality and more.
Catalog No. 50-008-28057 Supplier Enterprise Technology Solutions Supplier No. HOSVPF238
Vinyl Powder Free Exam Gloves provide a latex-free and economical solution to hand protection. Formulated from a special synthetic component and soft, PVC material, these gloves are soft like premium leather and fit snugly and comfortably on your hands. They are 100 percent free of rubber latex, eliminating the risk of developing latex protein-related allergies. 4 mil thick, powder-free gloves are perfect for use in industries such as cosmetology, food-processing, government, grocery, healthcare, hospitality and more.