Learn More
Enterprise Technology Solutions Hospeco Latex Flock-Lined Gloves, 16 Mil, 12" Long, Small, 12/DZ, Yellow
SDP
Catalog No. 5000828078
Encompass_Preferred
Safety Zone latex, flock-lined "dishwashing gloves" provide dependable hand protection for general cleaning, dishwashing and food handling. Made from waterproof, flexible natural latex, they offer resistance to mild cleaning agents, water-based chemicals, acids, fats, alkalis, alcohol, acetone and some solvents. These 16 mil, 12" long gloves help resist tears and punctures. Soft, cotton flock lining absorbs sweat for all-day comfort. Recessed diamond texture ensures a secure, nonslip grip. Rolled cuffs help divert water and allow easy on/off. Cost-effective and flexible, these size small gloves deliver excellent touch sensitivity and dexterity for janitorial, warewashing, food processing, light chemical handling and other general cleaning tasks.
Catalog No. 50-008-28078 Supplier Enterprise Technology Solutions Supplier No. HOSGRFYSM1B
Safety Zone latex, flock-lined "dishwashing gloves" provide dependable hand protection for general cleaning, dishwashing and food handling. Made from waterproof, flexible natural latex, they offer resistance to mild cleaning agents, water-based chemicals, acids, fats, alkalis, alcohol, acetone and some solvents. These 16 mil, 12" long gloves help resist tears and punctures. Soft, cotton flock lining absorbs sweat for all-day comfort. Recessed diamond texture ensures a secure, nonslip grip. Rolled cuffs help divert water and allow easy on/off. Cost-effective and flexible, these size small gloves deliver excellent touch sensitivity and dexterity for janitorial, warewashing, food processing, light chemical handling and other general cleaning tasks.