Learn More
Enterprise Technology Solutions Hospeco TriggerSprayer,1.4mLOutpt,3FinGrip,28/400mmNeck,250/CT,BE/WE
SDP
Catalog No. 5000811313
Encompass_Preferred
General Purpose Trigger Sprayer features a three-finger grip to reduce hand fatigue when handling most any project. Two-piston design allows better chemical resistance. Sprayer tube measures 10" and connects to a 28/400mm neck finish. Trigger spray produces a 1.4mL output.
Catalog No. 50-008-11313 Supplier Enterprise Technology Solutions Supplier No. HOS4902
General Purpose Trigger Sprayer features a three-finger grip to reduce hand fatigue when handling most any project. Two-piston design allows better chemical resistance. Sprayer tube measures 10" and connects to a 28/400mm neck finish. Trigger spray produces a 1.4mL output.