Learn More
Sklar Instruments Hospital Bucket, Sklar, Stainless Steel. This product has a diameter of 11-3/4 in. and height of 10-1/8 in. with a 16 quart capacity. A lid is not included. EA/1.
SDP
Catalog No. 501436946 Shop All Sklar Instruments Products
Encompass_Preferred
Hospital Bucket, Sklar, Stainless Steel. Sklar's Stainless Steel Hospital Bucket may serve a variety of purposes but are most commonly used for collecting liquids or dressings until they can be properly disposed of. This product has a diameter of 11-3/4 in. and height of 10-1/8 in. with a 16 quart capacity. A lid is not included.
This item is not returnable. View return policy
Catalog No. 50-143-6946 Supplier Sklar Instruments Supplier No. 101183
Hospital Bucket, Sklar, Stainless Steel. Sklar's Stainless Steel Hospital Bucket may serve a variety of purposes but are most commonly used for collecting liquids or dressings until they can be properly disposed of. This product has a diameter of 11-3/4 in. and height of 10-1/8 in. with a 16 quart capacity. A lid is not included.