Learn More
MSC Rochester Midland Disinfectant, Hospital Type, Ready to Use, 6x1 qt/case
Catalog No. 502838789
Encompass
This ready-to-use, hospital-type disinfectant, cleaner, and deodorizer is formulated for use on hard, non-porous inanimate surfaces. It is effective against a broad spectrum of pathogens, including SARS-CoV-2, Tuberculosis, Norovirus, HIV (Aids), and Canine Parvovirus, offering fast kill times.
  • Ready to use formulation for convenience.
  • Effective against a wide range of pathogens including SARS-CoV-2, Tuberculosis, Norovirus, HIV (Aids), and Canine Parvovirus.
  • Eliminates odors and leaves a refreshing lemon scent.
  • Kills 99.9% of bacteria.
  • Cleans, disinfects, and deodorizes in one simple step.
  • Cost effective and time efficient when used with a sprayer.
  • Features a clear amber color and a pH of 12.5.
  • Triggers are not included.
Catalog No. 50-283-8789 Supplier MSC Supplier No. 63822654
This ready-to-use, hospital-type disinfectant, cleaner, and deodorizer is formulated for use on hard, non-porous inanimate surfaces. It is effective against a broad spectrum of pathogens, including SARS-CoV-2, Tuberculosis, Norovirus, HIV (Aids), and Canine Parvovirus, offering fast kill times.
  • Ready to use formulation for convenience.
  • Effective against a wide range of pathogens including SARS-CoV-2, Tuberculosis, Norovirus, HIV (Aids), and Canine Parvovirus.
  • Eliminates odors and leaves a refreshing lemon scent.
  • Kills 99.9% of bacteria.
  • Cleans, disinfects, and deodorizes in one simple step.
  • Cost effective and time efficient when used with a sprayer.
  • Features a clear amber color and a pH of 12.5.
  • Triggers are not included.