Learn More
Alpco Ltd Hospital Stool Panel RT-PCR Kit
SDP
Catalog No. NC2284606
Encompass
Hospital Stool Panel is a multiplex real-time RT-PCR kit for the direct, qualitative detection and differentiation of norovirus (genogroup I and II), rotavirus and Clostridium difficile toxin genes A (tcdA) / B (tcdB) in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.
This item is not returnable. View return policy
Catalog No. NC2284606 Supplier Alpco Ltd Supplier No. 86HSPHU100
Hospital Stool Panel is a multiplex real-time RT-PCR kit for the direct, qualitative detection and differentiation of norovirus (genogroup I and II), rotavirus and Clostridium difficile toxin genes A (tcdA) / B (tcdB) in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.