Learn More
MSC Clorox Healthcare Disinfectant and Deodorizer, Citrus, 14 oz Aerosol, 12/Case
Catalog No. 502869877
Encompass
This all-purpose disinfectant and deodorizer is designed for use in healthcare settings. It kills a broad spectrum of microorganisms on hard, non-porous surfaces and soft surfaces, including bacteria, viruses, and fungi. With a light citrus scent, it also sanitizes the air by eliminating odor-causing bacteria and prevents mold and mildew growth on fabrics.
  • Kills 24 types of microorganisms on hard surfaces in 5 minutes
  • Effective against SARS-CoV-2, the virus that causes COVID-19
  • Kills 99.9% of bacteria on soft surfaces in 30 seconds
  • Eliminates odors and freshens air instantly with citrus scent
  • Prevents mold and mildew growth on fabrics
  • Versatile for use on hard, non-porous surfaces and fabrics
  • Suitable for hospitals, medical facilities, and veterinary offices
Catalog No. 50-286-9877 Supplier MSC Supplier No. 91482521
This all-purpose disinfectant and deodorizer is designed for use in healthcare settings. It kills a broad spectrum of microorganisms on hard, non-porous surfaces and soft surfaces, including bacteria, viruses, and fungi. With a light citrus scent, it also sanitizes the air by eliminating odor-causing bacteria and prevents mold and mildew growth on fabrics.
  • Kills 24 types of microorganisms on hard surfaces in 5 minutes
  • Effective against SARS-CoV-2, the virus that causes COVID-19
  • Kills 99.9% of bacteria on soft surfaces in 30 seconds
  • Eliminates odors and freshens air instantly with citrus scent
  • Prevents mold and mildew growth on fabrics
  • Versatile for use on hard, non-porous surfaces and fabrics
  • Suitable for hospitals, medical facilities, and veterinary offices