Learn More
Cayman Chemical Host Cel ProteIns - HCPE 96Wel
SDP
Catalog No. 502984367
Encompass_Preferred
A selective nonpeptide CCR1 antagonist that can displace the endogenous CCR1 ligands MIP-1a RANTES and MCP-3 with Ki values of 1 2 8 and 5 5 nM respectively reduces chemotaxis of inflammatory cells in models of multiple sclerosis sepsis rheumatoid arthritis and lupus nephritis
Catalog No. 50-298-4367 Supplier Cayman Chemical Supplier No. 1891996 WELL
A selective nonpeptide CCR1 antagonist that can displace the endogenous CCR1 ligands MIP-1a RANTES and MCP-3 with Ki values of 1 2 8 and 5 5 nM respectively reduces chemotaxis of inflammatory cells in models of multiple sclerosis sepsis rheumatoid arthritis and lupus nephritis