Learn More
Zymo Research Corporation Host Depletion Solution, 20 ml
SDP
Catalog No. 501957587
Encompass_Preferred
Host Depletion Solution selectively lyses eukaryotic cells to reduce the amount of contaminating host DNA in a biological sample.
Catalog No. 50-195-7587 Supplier Zymo Research Corporation Supplier No. D4310120
Host Depletion Solution selectively lyses eukaryotic cells to reduce the amount of contaminating host DNA in a biological sample.