Learn More
Bioworld Hosta Initiation/Multiplication Medium Stage I/II, 50 L
Catalog No. 50254907
Encompass_Preferred
Hosta Initiation/Multiplication Medium Stage I/IIContains Macro and Micronutrients as described by Murashige & Skoog (1962) and the Vitamins as described by Linsmaier and Skoog (1965)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 43.39 g/LPartially soluble in water
Catalog No. 50-254-907 Supplier Bioworld Supplier No. 306301654
Hosta Initiation/Multiplication Medium Stage I/IIContains Macro and Micronutrients as described by Murashige & Skoog (1962) and the Vitamins as described by Linsmaier and Skoog (1965)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 43.39 g/LPartially soluble in water