Learn More
Zymo Research Corporation HostZERO Microbial DNA Kit (50 preps)
SDP
Catalog No. NC1621465
Encompass_Preferred
This kit uses a novel method to reduce the amount of contaminating host DNA by selectively lysing the eukaryotic cells and degrading this DNA prior to total DNA purification. Paired with Zymo Researchs non-biased purification technology, the HostZERO™ Microbial DNA Kit allows for the exclusive capture of DNA from living microbial cells in a biological sample.This new technology is able to reduce the presence of human DNA in a saliva sample from 65% (untreated sample) to less than 1% (treated sample, Figure 1). Concurrently, the depletion process utilizes the ZymoBIOMICS™ non-biased DNA isolation technology for accurate data representation (Figure 2). Finally, the HostZERO™ Microbial DNA Kit recovers the highest amount of bacterial DNA as compared to other methods (Figure 3). By removing the presence of host DNA and reducing bias in purification, the HostZERO™ Microbial DNA Kit produces the highest-quality data for microbial samples.
Catalog No. NC1621465 Supplier Zymo Research Corporation Supplier No. D4310
This kit uses a novel method to reduce the amount of contaminating host DNA by selectively lysing the eukaryotic cells and degrading this DNA prior to total DNA purification. Paired with Zymo Researchs non-biased purification technology, the HostZERO™ Microbial DNA Kit allows for the exclusive capture of DNA from living microbial cells in a biological sample.This new technology is able to reduce the presence of human DNA in a saliva sample from 65% (untreated sample) to less than 1% (treated sample, Figure 1). Concurrently, the depletion process utilizes the ZymoBIOMICS™ non-biased DNA isolation technology for accurate data representation (Figure 2). Finally, the HostZERO™ Microbial DNA Kit recovers the highest amount of bacterial DNA as compared to other methods (Figure 3). By removing the presence of host DNA and reducing bias in purification, the HostZERO™ Microbial DNA Kit produces the highest-quality data for microbial samples.