Learn More
Apexbio Technology LLC HOSu 6066-82-6 250g
SDP
Catalog No. 5000061875
Encompass_Preferred
HOSu (N-hydroxysuccinimide) is a reactive ester reagent widely utilized in biochemical and biomedical research Chemically it functions by activating carboxyl groups to form a transient ester intermediate allowing subsequent formation of amide bonds through nucleophilic substitution between the ester intermediate and suitable amino-containing biomolecules This reaction mechanism provides a straightforward approach for controlled conjugation of proteins peptides or other amine-bearing molecules Typical research applications include protein labeling peptide conjugation functionalization of biosensor surfaces and bioconjugation of biomolecules onto nanoparticle carriers or biomaterials
Catalog No. 50-000-61875 Supplier Apexbio Technology LLC Supplier No. A7059250000
HOSu (N-hydroxysuccinimide) is a reactive ester reagent widely utilized in biochemical and biomedical research Chemically it functions by activating carboxyl groups to form a transient ester intermediate allowing subsequent formation of amide bonds through nucleophilic substitution between the ester intermediate and suitable amino-containing biomolecules This reaction mechanism provides a straightforward approach for controlled conjugation of proteins peptides or other amine-bearing molecules Typical research applications include protein labeling peptide conjugation functionalization of biosensor surfaces and bioconjugation of biomolecules onto nanoparticle carriers or biomaterials