Learn More
Fine Science Tools Hot Bead Sterilizers, 20.5 x 12.5 x 13.5 cm
SDP
Catalog No. NC9779311
Encompass
This sterilizer provides a safe, effective and convenient method for sterilizing your surgical instruments prior to surgery or tissue culture. Without using potentially dangerous gases, flames or chemicals, you can ensure that your instruments will be free of pathogens and microbial contaminants. The beads take 15-20 minutes to reach 240C to 270C (464F to 518F), and then the sterilizer is ready for use. The temperature is thermostatically maintained, and the unit can be left on all day. Instruments require approximately 20 seconds of contact with the beads for complete sterilization. The sterilizer is safe to use even in laminar flow hoods. Instruments not included. Product: Hot Bead Sterilizer - 14cm Well/ Dimensions: 20.5 x 12.5 x 13.5cm/ Well Depth: 14cm/ Well Diameter: 4cm/ Well Volume: 175cm3 Unit: Each
Catalog No. NC9779311 Supplier Fine Science Tools Supplier No. 1800050
This sterilizer provides a safe, effective and convenient method for sterilizing your surgical instruments prior to surgery or tissue culture. Without using potentially dangerous gases, flames or chemicals, you can ensure that your instruments will be free of pathogens and microbial contaminants. The beads take 15-20 minutes to reach 240C to 270C (464F to 518F), and then the sterilizer is ready for use. The temperature is thermostatically maintained, and the unit can be left on all day. Instruments require approximately 20 seconds of contact with the beads for complete sterilization. The sterilizer is safe to use even in laminar flow hoods. Instruments not included. Product: Hot Bead Sterilizer - 14cm Well/ Dimensions: 20.5 x 12.5 x 13.5cm/ Well Depth: 14cm/ Well Diameter: 4cm/ Well Volume: 175cm3 Unit: Each