Learn More
Cole-Parmer Environmental Express HotBlock SC100 Digestion System, 36-Well, 50 mL; 120 VAC
Catalog No. NC1775612 Shop All Cole-Parmer Products
Encompass
EPA-approved systems that simplify and improve metals prep and digestions. Blocks are PTFE coated for chemical and corrosion resistance. Integrated block heater and controller design. Set high and low temperature alarm limits. Include polycarbonate transfer racks designed to set on the block.
Catalog No. NC1775612 Supplier Cole-Parmer Supplier No. 5320502
EPA-approved systems that simplify and improve metals prep and digestions. Blocks are PTFE coated for chemical and corrosion resistance. Integrated block heater and controller design. Set high and low temperature alarm limits. Include polycarbonate transfer racks designed to set on the block.