Learn More
Bullard™ Hot/Cold Control Tube Assemblies for Body Temperature Management Devices, Quick-Disconnect Nipples
SDP
Catalog No. 19400004 Shop All Bullard Company Products
Change view
Click to view available options
Material:
Brass
Stainless Steel
Steel
Connection Type:
Bayonet
CEJN
Schrader
Snap-Tite
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Connection Type
19-400-004 Stainless Steel CEJN
19-400-002 Steel Snap-Tite
19-400-003 Brass Snap-Tite
19-400-001 Steel Schrader
19-400-005 Steel Bayonet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-400-004 Supplier Bullard™ Supplier No. HC240037
Add to Cart
Add to Cart
Hot/cold tube assemblies are for use with GenVX, 88VX, CC20, and GR50 series airline respirators.
  • Supplied-Air Respirator (SAR) body temperature management accessory
  • Hot/cold adjustable tubes DC50
  • 0.25 in. coupling
  • Choose from a variety of materials and fastenings
Specifications
Certifications/Compliance CE, NIOSH
Connection Type CEJN
Material Stainless Steel
Product Type Hose Nipple
For Use With (Equipment) GenVX, 88VX, CC20, RT and GR50 series airline respirator
Includes Heat shield
Model HC240037
Diameter (English) Interior 0.25 in.
Diameter (Metric) Interior 0.63 cm
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.