Learn More
Chemglass Life Sciences Hot/Cold Plate for 135 mm Hot Plate Stirrers and 135 mm Reaction Blocks
SDP
Catalog No. NC3866787
Encompass_Preferred
Hot/cold plate assembly designed for 135 mm diameter hot plate stirrers and reaction blocks. It is configured for connection to an external temperature control unit to provide heating or cooling and includes inlet/outlet ports and a condensation collection tray with a drain fitting. Hot plates, reaction blocks, control unit, and tubing are sold separately.
  • Designed for 135 mm diameter hot plate stirrers and reaction blocks.
  • Provides heated or chilled circulation when connected to an external temperature control unit.
  • M16x1 male inlet and outlet connections for temperature control circulation.
  • Condensation collection tray with 1/4 in OD compression fitting for drainage.
  • Requires a separate temperature control unit; accessories sold separately.
  • Machined product line construction.
Catalog No. NC3866787 Supplier Chemglass Life Sciences Supplier No. CG1991C200
Hot/cold plate assembly designed for 135 mm diameter hot plate stirrers and reaction blocks. It is configured for connection to an external temperature control unit to provide heating or cooling and includes inlet/outlet ports and a condensation collection tray with a drain fitting. Hot plates, reaction blocks, control unit, and tubing are sold separately.
  • Designed for 135 mm diameter hot plate stirrers and reaction blocks.
  • Provides heated or chilled circulation when connected to an external temperature control unit.
  • M16x1 male inlet and outlet connections for temperature control circulation.
  • Condensation collection tray with 1/4 in OD compression fitting for drainage.
  • Requires a separate temperature control unit; accessories sold separately.
  • Machined product line construction.