Learn More
Mayflower Bioscience HOT FIREPol DNA Polymerase Kit
SDP
Catalog No. 502963912
Encompass
Hot-start DNA Polymerase with a unique 30-day stability at room temperature for your everyday PCR needs. A chemically modified hot-start version of the thermostable Taq DNA polymerase FIREPol. This enzyme is activated only by pre-incubation at 95°C, preventing any unspecific polymerase activity at lower temperatures during reaction set-up.
Catalog No. 50-296-3912 Supplier Mayflower Bioscience Supplier No. 0102KIT00500
Hot-start DNA Polymerase with a unique 30-day stability at room temperature for your everyday PCR needs. A chemically modified hot-start version of the thermostable Taq DNA polymerase FIREPol. This enzyme is activated only by pre-incubation at 95°C, preventing any unspecific polymerase activity at lower temperatures during reaction set-up.