Learn More
Thermo Scientific™ Hot Gas Bypass Temperature Controllers
GSA_VA
Catalog No. TF195021 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Factory-Installed
Voltage:
120 V
Catalog No. Type Voltage
TF195021 Factory-Installed 120 V
1 options
Catalog No. TF195021 Supplier Thermo Scientific™ Supplier No. 195021
Add to Cart
Add to Cart
Combine these hot gas bypass temperature controllers with Thermo Scientific™ CryoPlus™ LN2 cryopreservation storage systems for increased utility.
Combine Thermo Scientific™ Hot Gas Bypass Temperature Controllers with Thermo Scientific™ CryoPlus™ LN2 cryopreservation storage systems for increased utility.
Features:
  • 120V or 220V
  • Factory or field installed
Ordering Information:
Field installed option must be installed by a qualified professional.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type Factory-Installed
For Use With (Equipment) CryoPlus 1,2,3,4
Voltage 120 V