Learn More
Electron Microscopy Sciences Hot Hand Protector Pad, 1 each
SDP
Catalog No. 5026588
Encompass_Preferred
Made from strong silicone and it withstands temperatures as high as 500 degree F (260 degree C). A gripping surface with built in multi suction studs for a non-slip grip. Has two pockets for thumb and fingers.
This item is not returnable. View return policy
Catalog No. 50-265-88 Supplier Electron Microscopy Sciences Supplier No. 64480
Made from strong silicone and it withstands temperatures as high as 500 degree F (260 degree C). A gripping surface with built in multi suction studs for a non-slip grip. Has two pockets for thumb and fingers.