Learn More
Research Products International Corp Hot Hand Protector, Red Silicone Rubber
SDP
Catalog No. 50550405
Encompass
Hot Hand Protector is molded of silicone rubber to provide superior protection for handling hot or cold objects. The material withstands temperatures up to 500°F (260°C) or down to -70°F (-57°C). Gripping surfaces have multiple studs with concave tips that increase grip strength. Highly visible color and hole for hanging keep them instantly available.
This item is not returnable. View return policy
Catalog No. 50-550-405 Supplier Research Products International Corp Supplier No. F38000
Hot Hand Protector is molded of silicone rubber to provide superior protection for handling hot or cold objects. The material withstands temperatures up to 500°F (260°C) or down to -70°F (-57°C). Gripping surfaces have multiple studs with concave tips that increase grip strength. Highly visible color and hole for hanging keep them instantly available.