Learn More
Grainger Transparent Hot Melt Glue Stick, 29/64" Diameter, 4" Length, 30/pk
Catalog No. NC1781922
Encompass_Preferred
Hot melt glue stick is made to be used as an adhesive when melted in an electric glue gun. The hot melt adhesive can be an alternative to solvent-based adhesives.
Catalog No. NC1781922 Supplier Grainger Supplier No. 23X756
Hot melt glue stick is made to be used as an adhesive when melted in an electric glue gun. The hot melt adhesive can be an alternative to solvent-based adhesives.