Learn More
Electron Microscopy Sciences Clear Thermoplastic Hot Mount Resin 1 KG
SDP
Catalog No. 50980275
Encompass_Preferred
Clear acryl, thermoplastic. Transparent mounts. Porous specimens. Surface electrical insulator for ConduFast.
Catalog No. 50-980-275 Supplier Electron Microscopy Sciences Supplier No. 1212
Clear acryl, thermoplastic. Transparent mounts. Porous specimens. Surface electrical insulator for ConduFast.