Learn More
Andwin Scientific HOT NOT GLOVE/NOMEX III 1PR
SDP
Catalog No. NC9025182
Encompass_Preferred
The Heat Resistant Gloves are able to protect users from 700 Degrees Farenheit. The gloves are anti-static. This product comes in one size only.
This item is not returnable. View return policy
Catalog No. NC9025182 Supplier Andwin Scientific Supplier No. 23217
The Heat Resistant Gloves are able to protect users from 700 Degrees Farenheit. The gloves are anti-static. This product comes in one size only.