Learn More
Hach Company Hot plate, 7x7 in., digital, 115 Vac
Supplier:  Hach Company 2881500
Encompass
Easy-to-read digital temperature settings features a flashing warning(CAUTION! HOT TOP) when the surface exceeds 50°C. Low-profiledesign to improve safety. Reflective, white ceramic top issmooth, seamless and resistant to alkali/acid solutions. Detachablecord. Built-in support rod holder (order support rod separately).Operating temperature range: 5 to 540°C (41 to 1004°F).
Catalog No. 50-199-6763