Learn More
World Precision Instrument Hot Plate Analgesia Meter
SDP
Catalog No. NC2335577
Encompass
Latency testing in rodents
To use the Hot Plate Analgesia meters, simply place the animals on a black anodized, aluminum plate (11 x 10.5 x 0.75”, 275 x 263 x 15 mm) and set the plate’s surface temperature to the desired set point (up to 75˚C). The plate maintains a consistent temperature throughout the test.
Catalog No. NC2335577 Supplier World Precision Instrument Supplier No. II39
Latency testing in rodents
To use the Hot Plate Analgesia meters, simply place the animals on a black anodized, aluminum plate (11 x 10.5 x 0.75”, 275 x 263 x 15 mm) and set the plate’s surface temperature to the desired set point (up to 75˚C). The plate maintains a consistent temperature throughout the test.