Learn More
Electron Microscopy Sciences SUPER-NUOVA™ Hot Plate Hot Plate Ceramic Top, 120 V
SDP
Catalog No. 501899818
Encompass_Preferred
Digital hote plate features a rugged low profile cast aluminum body provides stability and durability and a warning system that deters accidental contact with hot surface. The Remote probe allow for accurate reading of solution temperature to 0.1°C for temperature below 100°C. A user-adjustable over-temperature protection prevents runaway temperature conditions. Seamless reflective easy-to-clean, alkali/acid resistant white ceramic top.
Surface: 7.25"x7.25"
Catalog No. 50-189-9818 Supplier Electron Microscopy Sciences Supplier No. 7234212
Digital hote plate features a rugged low profile cast aluminum body provides stability and durability and a warning system that deters accidental contact with hot surface. The Remote probe allow for accurate reading of solution temperature to 0.1°C for temperature below 100°C. A user-adjustable over-temperature protection prevents runaway temperature conditions. Seamless reflective easy-to-clean, alkali/acid resistant white ceramic top.
Surface: 7.25"x7.25"