Learn More
Electron Microscopy Sciences SUPER-NUOVA™ Hot Plate Ceramic Top, 220 V
SDP
Catalog No. 501899830
Encompass_Preferred
Digital hote plate features a rugged low profile cast aluminum body provides stability and durability and a warning system that deters accidental contact with hot surface. The Remote probe allow for accurate reading of solution temperature to 0.1°C for temperature below 100°C. A user-adjustable over-temperature protection prevents runaway temperature conditions. Seamless reflective easy-to-clean, alkali/acid resistant white ceramic top.
Surface: 10.5"x10.5"
Catalog No. 50-189-9830 Supplier Electron Microscopy Sciences Supplier No. 7234520
Add to Cart
Add to Cart
Digital hote plate features a rugged low profile cast aluminum body provides stability and durability and a warning system that deters accidental contact with hot surface. The Remote probe allow for accurate reading of solution temperature to 0.1°C for temperature below 100°C. A user-adjustable over-temperature protection prevents runaway temperature conditions. Seamless reflective easy-to-clean, alkali/acid resistant white ceramic top.
Surface: 10.5"x10.5"