Learn More
Electron Microscopy Sciences Nuova™ Low End Hot Plates, 220/240 V, 10.25" x 10.25" heating surface, 540°C max range
SDP
Catalog No. 501899853
Encompass_Preferred
This hot plate features a corrosion resistant heating surface, low profile, & 24 month warranty. Durable & long lasting w/ a die cast aluminum case. Corrosion resistant w/ a porcelain coated stainless steel top. The topside drip edge protects internal components in case of spillage. Integral ring stand holder accomodates 0.5" (13 mm) support rod. Accomodates up to 20 lb (9.1 kg) loads. Recommended for use w/ glass vessels only. All unit included "PTFE" coated stir bar. Demand-Type thermostatic temperature control giving it excellent temperature stability +/-5.0 degree C (9 degF) at 371 °C (700 degF). Embedded heating elements – uniformity transfer heat to the top plate.Perfect choice for warming applications w/ low temperature control of 35 °C.All units operating temperature range is 38 – 371 degree C (100 –700 °F).Heating surface: 10.25" x 10.25"Range: 540°C maxAmps: 3.5Wattage: 840Footprint: 16.2 x 11.3"
Catalog No. 50-189-9853 Supplier Electron Microscopy Sciences Supplier No. 7235822
This hot plate features a corrosion resistant heating surface, low profile, & 24 month warranty. Durable & long lasting w/ a die cast aluminum case. Corrosion resistant w/ a porcelain coated stainless steel top. The topside drip edge protects internal components in case of spillage. Integral ring stand holder accomodates 0.5" (13 mm) support rod. Accomodates up to 20 lb (9.1 kg) loads. Recommended for use w/ glass vessels only. All unit included "PTFE" coated stir bar. Demand-Type thermostatic temperature control giving it excellent temperature stability +/-5.0 degree C (9 degF) at 371 °C (700 degF). Embedded heating elements – uniformity transfer heat to the top plate.Perfect choice for warming applications w/ low temperature control of 35 °C.All units operating temperature range is 38 – 371 degree C (100 –700 °F).Heating surface: 10.25" x 10.25"Range: 540°C maxAmps: 3.5Wattage: 840Footprint: 16.2 x 11.3"