Learn More
Research Products International Corp Digital Hot Plate Stirrer, 12 x 12 Inch Ceramic Top
SDP
Catalog No. 50999044
Encompass
The EchoTherm HS51 is designed to be the easiest to use and most accurate fully featured stirring hot plates in the world. Temperature measurement of a solution is accurate to 1% over the entire temperature range using a 100 ohm platinum RTD sensor and measurement system. Calibration of temperature is electronic and stored in memory. The calibration is set at the factory but can be done easily in the field using the probe calibration kit offered as an accessory. The chassis on both models is cast aluminum designed to keep spills and heat out of the electronics. They are coated with chemically resistant epoxy based paint for durability.
Catalog No. 50-999-044 Supplier Research Products International Corp Supplier No. 150610
The EchoTherm HS51 is designed to be the easiest to use and most accurate fully featured stirring hot plates in the world. Temperature measurement of a solution is accurate to 1% over the entire temperature range using a 100 ohm platinum RTD sensor and measurement system. Calibration of temperature is electronic and stored in memory. The calibration is set at the factory but can be done easily in the field using the probe calibration kit offered as an accessory. The chassis on both models is cast aluminum designed to keep spills and heat out of the electronics. They are coated with chemically resistant epoxy based paint for durability.