Learn More
Biocare Medical, LLC Hot Rinse, 25X - 500 ml
SDP
Catalog No. 5082889
Encompass_Preferred
Hot Rinse, 25X is intended for laboratory use in immunohistochemistry procedures on formalin-fixed paraffin-embedded (FFPE) tissues to ensure complete deparaffinization. Hot Rinse is sodium azide and thimerosal free, non-toxic, non-flammable and odorless. After deparaffinizing tissues in Reveal, Universal or Borg Decloaker, a subsequent hot rinse ensures complete deparaffinization. It is specially designed for use with Biocare Decloaking Chamber and is also compatible with other commercial heat retrieval deparaffinization solutions.
Catalog No. 50-828-89 Supplier Biocare Medical, LLC Supplier No. HTR1001M
Hot Rinse, 25X is intended for laboratory use in immunohistochemistry procedures on formalin-fixed paraffin-embedded (FFPE) tissues to ensure complete deparaffinization. Hot Rinse is sodium azide and thimerosal free, non-toxic, non-flammable and odorless. After deparaffinizing tissues in Reveal, Universal or Borg Decloaker, a subsequent hot rinse ensures complete deparaffinization. It is specially designed for use with Biocare Decloaking Chamber and is also compatible with other commercial heat retrieval deparaffinization solutions.