Learn More
Bellco Glass, Inc. Hot Shaker Flask Tray,125mL For Single Pan Units Only
SDP
Catalog No. 501946167
Encompass_Preferred
Stainless steel tray measures 9.75" × 17.75" and includes 10 stainless steel clamps for 125ml Erlenmeyer flasks. Stainless steel Measures: 9.75" deep ×17.75" wide (approx.) For single pan only. MADE IN THE USA
Catalog No. 50-194-6167 Supplier Bellco Glass, Inc. Supplier No. 774610125
Stainless steel tray measures 9.75" × 17.75" and includes 10 stainless steel clamps for 125ml Erlenmeyer flasks. Stainless steel Measures: 9.75" deep ×17.75" wide (approx.) For single pan only. MADE IN THE USA