Learn More
Bellco Glass, Inc. Hot Shaker Flask Tray,250mL For Single Pan Units Only
SDP
Catalog No. 501946168
Encompass_Preferred
Hot Shaker Flask Clamp Tray Use with Single Pan Hot Shaker Only. Tray fits inside the SST pan has rubber bumpers for a snug fit. Holds eight (8) 250ml shake flasks, two rows of 4. Material of Tray: SST .031" thick w/electropolish finishDimensions: 9 3/4" d x 17 3/4" wide (approx). MADE IN THE USA
Catalog No. 50-194-6168 Supplier Bellco Glass, Inc. Supplier No. 774610250
Hot Shaker Flask Clamp Tray Use with Single Pan Hot Shaker Only. Tray fits inside the SST pan has rubber bumpers for a snug fit. Holds eight (8) 250ml shake flasks, two rows of 4. Material of Tray: SST .031" thick w/electropolish finishDimensions: 9 3/4" d x 17 3/4" wide (approx). MADE IN THE USA