Learn More
Bellco Glass, Inc. Hot Shaker Flask Tray,500mL For Single Pan Units Only
SDP
Catalog No. 501946169
Encompass_Preferred
Hot Shaker Flask Clamp Tray Use with Single Pan Hot Shaker Only. Tray fits inside the SST pan has rubber bumpers for a snug fit. Holds six (6) 500mL Shake Flasks, 2 rows of 3. Material of Tray: SST .031" thick with electropolish finish. MADE IN THE USA
Catalog No. 50-194-6169 Supplier Bellco Glass, Inc. Supplier No. 774610500
Hot Shaker Flask Clamp Tray Use with Single Pan Hot Shaker Only. Tray fits inside the SST pan has rubber bumpers for a snug fit. Holds six (6) 500mL Shake Flasks, 2 rows of 3. Material of Tray: SST .031" thick with electropolish finish. MADE IN THE USA