Learn More
Bellco Glass, Inc. Hot Shaker Plus,230V
SDP
Catalog No. 501946172
Encompass_Preferred
The Bellco Sci-ERA Hot Shaker is a slow speed, large orbit shaker for incubation of electrophoretic gels and filter hybridization media. Stringent temperature control with a sensitivity of +/-0.1˚C is useful for many biochemical assays. This Bellco design incorporates a seamless stainless steel lined temperature controlled water bath mounted on a low-profile one inch orbiting platform. The unique chamber design offers simple containment of potential spills. The chamber is completely isolated from electromechanical components. The Sci-ERA Hot Shaker has audible over-temperature alarms and built-in safety features to prevent heater burn-out. An easy-to-use microprocessor controller offers stringent temperature control.. MADE IN THE USA
Catalog No. 50-194-6172 Supplier Bellco Glass, Inc. Supplier No. 774622220
Add to Cart
Add to Cart
Request a Quote
The Bellco Sci-ERA Hot Shaker is a slow speed, large orbit shaker for incubation of electrophoretic gels and filter hybridization media. Stringent temperature control with a sensitivity of +/-0.1˚C is useful for many biochemical assays. This Bellco design incorporates a seamless stainless steel lined temperature controlled water bath mounted on a low-profile one inch orbiting platform. The unique chamber design offers simple containment of potential spills. The chamber is completely isolated from electromechanical components. The Sci-ERA Hot Shaker has audible over-temperature alarms and built-in safety features to prevent heater burn-out. An easy-to-use microprocessor controller offers stringent temperature control.. MADE IN THE USA