Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hot and Spicy Molecules Model Set
SCI_ED

Catalog No. S94029
Change view
Click to view available options
Hot and spicy molecules model set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S94029 Hot and spicy molecules model set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94029 Supplier Ryler Enterprises Supplier No. HOT1

missing translation for 'validMainframeMsg'

Includes

Kit Contents:

  • 65 white 1-peg hydrogen atom centers
  • 5 blue 3-peg nitrogen atom centers
  • 14 red 2-peg oxygen atom centers
  • 4 yellow 2-peg sulfur atom centers
  • 3 yellow 4-peg sulfur atom centers
  • 3 silver 6-peg sulfur atom centers
  • 40 black 4-peg carbon atom centers
  • 13 black 3-peg carbon atom centers
  • 138-1.25 in. PVC, clear bonds (single bonds)
  • 18-1.625 in. PVC, clear bonds (double bonds)

Great activity for junior high through college biology students. A listing of 31 spices and their constituent compounds is contained in the accompanying booklet. The molecular structures of 63 spicy compounds guide students in making models.

The molecular structures of 63 spicy compounds guide students in making models.

  • Color-coded, modeled plastic atom centers are 1/4 in. diameter
  • Pegs are 3/8 in. long and are set at angles peculiar to the elements the atoms represent
  • Standard color-coding
  • Atom centers are bonded by connecting pegs with PVC tubing

Specifications

For Use With (Application) Junior high through college biology students
Includes 65 White 1-peg Hydrogen atom centers, 5 blue 3-peg Nitrogen atom centers, 14 red 2-peg Oxygen atom centers, 4 yellow 2-peg Sulfur atom centers, 3 yellow 4-peg Sulfur atom centers, 3 silver 6-peg Sulfur atom centers, 40 black 4-peg Carbon atom centers, 13
Product Type Molecules Model Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.