Learn More
Research Products International Corp HotSpot Safety Mat, Silicone Rubber, Red, 10 x 10 Inches
SDP
Catalog No. 50998805
Encompass
HotSpot Safety Mat provides the ideal surface protection in the laboratory. Silicone rubber mat features a non-slip ribbed surface that holds glassware securely in place on flat or angled surfaces and ideal for use with orbital and rocking shakers. HotSpot Safety Mat cushions fragile glass items and protects surfaces from potential heat damage. Safety mat is also convenient to use in microwave oven.
Catalog No. 50-998-805 Supplier Research Products International Corp Supplier No. 100352
HotSpot Safety Mat provides the ideal surface protection in the laboratory. Silicone rubber mat features a non-slip ribbed surface that holds glassware securely in place on flat or angled surfaces and ideal for use with orbital and rocking shakers. HotSpot Safety Mat cushions fragile glass items and protects surfaces from potential heat damage. Safety mat is also convenient to use in microwave oven.