Learn More
Genscript Corporation Hot Start Taq DNA Polymerase
Supplier:  Genscript Corporation E000491000
Encompass
Hot Start Taq DNA Polymerase is a recombinant thermostable Taq DNA polymerase complexed with a thermolabile neutralizing antibody that blocks the polymerase activity prior to the initial DNA denaturation step of PCR When the temperature of the PCR reaction mix reaches 95 C during the initial DNA denaturing step of PCR cycling activity of the Taq DNA polymerase is fully restored br/
Catalog No. 50-000-58352