Learn More
Grainger Hot Surface Igniter OEM 120V AC
Catalog No. 502767066
Encompass_Preferred
The Honeywell Home Hot Surface Igniter is an original equipment manufacturer (OEM) replacement part that fits many brands of gas furnaces, including Carrier, Bryant, Payne, and others. It has a 120VAC voltage rating and measures approximately 3-1/2" L x 1-3/4" W x 1/2" H. This ceramic igniter replaces older style glow plug igniters and provides better durability and reliability. The igniter heats up to approximately 1800°F to ignite the gas burner in a gas furnace. It is designed for an easy, direct replacement install. The Honeywell igniter comes with a 1 year manufacturer's warranty.
Catalog No. 50-276-7066 Supplier Grainger Supplier No. 16X598
The Honeywell Home Hot Surface Igniter is an original equipment manufacturer (OEM) replacement part that fits many brands of gas furnaces, including Carrier, Bryant, Payne, and others. It has a 120VAC voltage rating and measures approximately 3-1/2" L x 1-3/4" W x 1/2" H. This ceramic igniter replaces older style glow plug igniters and provides better durability and reliability. The igniter heats up to approximately 1800°F to ignite the gas burner in a gas furnace. It is designed for an easy, direct replacement install. The Honeywell igniter comes with a 1 year manufacturer's warranty.