Learn More
Grainger Hot Surface Igniter Universal 102-132VAC
Catalog No. 502767135
Encompass_Preferred
The White Rodgers Hot Surface Igniter is a universal replacement igniter that fits a wide range of furnaces including Carrier, Lennox, York, Trane, and more. It has a 120VAC voltage rating and an amperage range of 3.0 to 4.5 amps. The ceramic construction provides durability. This is an OEM replacement part that measures 7.5 inches in length. It features a ceramic insulator to prevent shorting. The igniter has a 90 day warranty.
Catalog No. 50-276-7135 Supplier Grainger Supplier No. 16X597
The White Rodgers Hot Surface Igniter is a universal replacement igniter that fits a wide range of furnaces including Carrier, Lennox, York, Trane, and more. It has a 120VAC voltage rating and an amperage range of 3.0 to 4.5 amps. The ceramic construction provides durability. This is an OEM replacement part that measures 7.5 inches in length. It features a ceramic insulator to prevent shorting. The igniter has a 90 day warranty.