Learn More
MSC Belleville Assault Boot, 8 in., Steel Toe, Coyote Brown, 1 ea.
Catalog No. 5000686501
Encompass
The 533 ST is an 8 in. hot weather assault boot featuring a hybrid design with a protective steel toe. Engineered for performance in rugged environments, this boot combines durable construction with advanced features to maximize mobility and traction.
  • Constructed with 100% cattlehide leather and nylon fabric
  • Steel toe protection meets ASTM F2412-24 and F2413-24 standards with EH rating
  • Exclusive VIBRAM® Ibex rubber outsole with sharp angled lugs and defined heel
  • Highly cushioned polyurethane midsole for shock absorption
  • Breathable moisture-wicking lining
  • Deeply serrated toe bumper designed for vertical climbing and crawling
  • Aggressive lace-to-toe style for a customized fit
  • US Navy certified and Berry compliant
Catalog No. 50-006-86501 Supplier MSC Supplier No. 47592282
The 533 ST is an 8 in. hot weather assault boot featuring a hybrid design with a protective steel toe. Engineered for performance in rugged environments, this boot combines durable construction with advanced features to maximize mobility and traction.
  • Constructed with 100% cattlehide leather and nylon fabric
  • Steel toe protection meets ASTM F2412-24 and F2413-24 standards with EH rating
  • Exclusive VIBRAM® Ibex rubber outsole with sharp angled lugs and defined heel
  • Highly cushioned polyurethane midsole for shock absorption
  • Breathable moisture-wicking lining
  • Deeply serrated toe bumper designed for vertical climbing and crawling
  • Aggressive lace-to-toe style for a customized fit
  • US Navy certified and Berry compliant