Learn More
Cole-Parmer Environmental Express HotBlock SC154 Digestion System, 54-Well, 50 mL; 120 VAC
Catalog No. NC1139991 Shop All Cole-Parmer Products
Encompass_Preferred
This is an EPA-approved system that simplifies and improves metals prep and digestions. It features an integrated block heater and controller design. Blocks are PTFE coated for chemical and corrosion resistance. Users can set high and low temperature alarm limits. System includes polycarbonate transfer racks designed to set on the block.
Catalog No. NC1139991 Supplier Cole-Parmer Supplier No. 5320504
This is an EPA-approved system that simplifies and improves metals prep and digestions. It features an integrated block heater and controller design. Blocks are PTFE coated for chemical and corrosion resistance. Users can set high and low temperature alarm limits. System includes polycarbonate transfer racks designed to set on the block.