Learn More
MSC Hoto Instruments Durometer, Shore D, 0-100 Durometer, 1 Point Accuracy
Catalog No. 5000513651
Encompass
This portable dial hardness tester, the E2-D, is designed for precise hardness measurements using the Shore D scale. It provides an accurate reading within a range of 0 to 100 durometer, ensuring reliable results for various materials.
  • Accurate to 1 point
  • Shore D scale measurement
  • Measures hardness from 0 to 100 durometer
  • Includes a 3-point calibration certificate traceable to NIST
  • Comes with a hard plastic carrying case
Catalog No. 50-005-13651 Supplier MSC Supplier No. 64177967
This portable dial hardness tester, the E2-D, is designed for precise hardness measurements using the Shore D scale. It provides an accurate reading within a range of 0 to 100 durometer, ensuring reliable results for various materials.
  • Accurate to 1 point
  • Shore D scale measurement
  • Measures hardness from 0 to 100 durometer
  • Includes a 3-point calibration certificate traceable to NIST
  • Comes with a hard plastic carrying case