Learn More
Description
- Provide reliable day-in, day-out performance so your laboratory glass is always ready-to-go
- Two independently operated chambers with up to four loading levels for high volume washing
- All models come with removable top and bottom stainless-steel open racks to accommodate a variety of glassware
- Spindle rack ready levels accommodate direct injection spindle racks (sold separately) for thorough cleaning inside narrow-neck glassware
- Stainless steel baskets, insert racks and accessories sold separately to maximize space, improve efficiency, and protect specialty glassware
- Temperature selection to 70°C (158°F); can effectively dissolve stubborn oils
- DI rinse models ensure a spotless finish; easily connect to fixed or portable DI water systems
- Optional DI Booster Pump recommended when the incoming DI water source has a pressure of less than 18psi
- Delayed start feature allows washer to operate unattended during off-peak hours to reduce energy costs
- Designed for optimal quiet operation with noise buffering insulation
- Three filters protect pumps from debris; conveniently located for easy access
- Minimal, efficient water consumption
- Enhanced dual fan drying system for spotless results
- 304 stainless steel chamber and exterior casing ensures corrosion protection and long life
- 3 in. lockable casters allow easy, controlled mobility of unit
- UL 61010-1 and CSA 22.2 61010-1 certified by MET Labs
- Microprocessor control system features 6 cycle selections including a customizable timed option to ensure minimum energy consumption. All wash cycles have pre-wash, wash, pre-rinse and rinse cycles to prevent contamination.
- Rinse Only - Short tap water rinse cycle
- Normal Wash - Lower temperature (55°C) longer cycle with three (3) final rinse cycles
- Daily Wash - Standard temperature (65°C) with two (2) final rinse cycles
- Heavy Wash - Higher temperature (70°C) longer cycle with multiple pre-wash and final rinse cycles
- Heavy Wash Time Saver - Lower temperature (55°C) version of Heavy Wash with a shorter overall cycle time
- Timed Wash Programs - Standard temperature (65°C) in programmable 15 minute increments up to 2.5 hours
Specifications
Specifications
| Description | DI Rinse - Top Standard Rack; Bottom Spindle Rack Ready |
| Display | Digital |
| Material | Stainless Steel |
| Max. Pressure | 60 psi |
| Water Consumption | Per Chamber: Min: Tap 2.4 gal. (9L), DI 0.8 gal. (3L); Max: Tap 4 gal. (15L), DI 1.6 gal. (6L) |
| Certifications/Compliance | MET Labs |
| For Use With (Application) | Washing Lab Glassware |
| Temperature Setting | 55°-70°C (131°-158°F) |
| Depth (English) | 23.31 in. |
| Depth (Metric) | 59.2 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.