Learn More
Electron Microscopy Sciences EMS Digital Hotplate, 115 V
SDP
Catalog No. 501927835
Encompass_Preferred
EMS hotplates, stirrers and hotplate-stirrers feature an exceptionally durable, chemical resistant, white ceramic work surface. Their space-efficient design (7 x 10.5" footprint), makes them ideal for use on crowded benchtops and inside bio-hoods.
Advanced microprocessor controls allow quick, precise adjustment of speed and temperature. The large, backlit LCD display provides constant visualization of the speed and temperature while a safety LED indicates when the top plate is above 50 degree C. In addition, an optional probe is available for applications that require precise temperature contol. When connected, the probe provides direct feedback to the microprocessor, which continuously adjusts and maintains the selected temperature in the sample within ± 0.5 degree C.
Catalog No. 50-192-7835 Supplier Electron Microscopy Sciences Supplier No. 8703003
EMS hotplates, stirrers and hotplate-stirrers feature an exceptionally durable, chemical resistant, white ceramic work surface. Their space-efficient design (7 x 10.5" footprint), makes them ideal for use on crowded benchtops and inside bio-hoods.
Advanced microprocessor controls allow quick, precise adjustment of speed and temperature. The large, backlit LCD display provides constant visualization of the speed and temperature while a safety LED indicates when the top plate is above 50 degree C. In addition, an optional probe is available for applications that require precise temperature contol. When connected, the probe provides direct feedback to the microprocessor, which continuously adjusts and maintains the selected temperature in the sample within ± 0.5 degree C.