Learn More
Sigma Aldrich Fine Chemicals Biosciences HotSealTM sealing film for
Catalog No. 502901071
Encompass
Ideal Use PCR applicationsMaterial PolyesterGas Permeable NoUV Transparent NoTemp. Range -40C to 125CThickness 2.3 mil (0.0023 in.)
Catalog No. 50-290-1071 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z740250100EA
Ideal Use PCR applicationsMaterial PolyesterGas Permeable NoUV Transparent NoTemp. Range -40C to 125CThickness 2.3 mil (0.0023 in.)